Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibberellic Acid, 90%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
77-06-5
C19H22O6
346.38
MFCD00079329
IXORZMNAPKEEDV-MOBSEVHISA-N
gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs
91757643
CC12C(C=CC3(C1C(C45C3CCC(C4)(C(=C)C5)O)C(=O)O)OC2=O)O
77-06-5
C19H22O6
346.38
MFCD00079329
IXORZMNAPKEEDV-MOBSEVHISA-N
gibberellic acid,gibberellin a3,gibberellin,gibreskol,brellin,cekugib,grocel,gibberellin x,gibberellic acid ga3,gib-tabs
91757643
CC12C(C=CC3(C1C(C45C3CCC(C4)(C(=C)C5)O)C(=O)O)OC2=O)O
Specific Rotation Condition 82.50 (20.00°C c=10,ethanol)
Specific Rotation 82.5
Packaging Glass bottle
Loss on Drying 3% max. (100°C, 7 hrs) (vacuum)
Quantity 25g
Melting Point 227.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Merck Index 15, 4454
Solubility Information Solubility in water: 5g/L (20° c). Other solubilities: soluble in aq.sol. of sodium bicarbonate,soluble in methanol,ethanol,acetone and aq. sol.,of sodium acetate,moderately soluble in ethyl acetate- slightly,soluble in ether- insoluble in chl
Formula Weight 346.38
Physical Form Crystalline Powder
Percent Purity 90%
Chemical Name or Material Gibberellic acid
Show More Show Less

GHS P Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning