Learn More
Gibberellic acid, 98%, Thermo Scientific Chemicals
Description
Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 222°C (decomposition) |
Color | White |
Quantity | 500 mg |
Beilstein | 54346 |
Merck Index | 14,4419 |
Solubility Information | Slightly soluble in water. Soluble in alcohol,isopropyl alcohol,ethanol,acetone and chloroform. |
Optical Rotation | +78° (c=2 in methanol) |
Formula Weight | 346.37 |
Percent Purity | 98% |
Physical Form | Powder |
Show More |
Safety and Handling
Hazard Category | H319 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 201-001-0 |
RTECSNumber | LY8990000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.