Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human FGF-basic (FGF-2/bFGF) Recombinant Protein
Human FGF-basic (FGF-2/bFGF) Recombinant Protein
Supplier: Gibco™ 13256029
Description
Human FGF-basic (FGF-2/bFGF) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
bFGF (also known as fibroblast growth factor-basic, FGF-basic, FGF-β, FGF2, or heparin binding growth factor) is used in studies of angiogenesis, mitogenesis of fibroblasts, neurite outgrowth in PC-12 cells, receptor binding, and tyrosine protein kinase phosphorylation.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: proteins bioassayed with Gibco media
- High-purity, recombinant, bioactive protein: to help ensure growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; recombinant human bFGF specific activity is determined by thymidine incorporation into serum-starved Balb/c 3T3 cells
- Proven compatibility: Recombinant proteins are bioassayed with Gibcomedia
Specifications
Cell Culture | |
10 μg | |
Human | |
Carrier-Free | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis |
PAGE | |
FGF-Basic | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
E. coli | |
Full Length, Recombinant, Ligand | |
FGFs (Fibroblast Growth Factors) | |
95 % |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction