Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ DMEM, High Glucose, HEPES Non-distribution product as customer accommodation.

Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, HEPES is used in cell culture processing applications.

Manufacturer:  Gibco™ 12430120

 View more versions of this product

Catalog No. 12-430-120


  Check Availability
Add to cart

Disclaimers:

When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco™ DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference their Type II Drug Master File (DMF). For human ex vivo tissue and cell culture processing applications.


Requires : Supplementation

Specifications

Description

  • Contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium
  • Originally formulated with low glucose (1g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (3.7g/L), and therefore requires a 5 to 10% CO2 environment to maintain physiological pH
  • Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, cell lines such as HeLa, 293, Cos-7, and PC-12
Description & Specifications

Specifications

Antibody and cell culture production, Mammalian cell culture
Calcium Chloride (CaCl2) (anhyd.), Choline chloride, D-Calcium pantothenate, D-Glucose (Dextrose), Ferric Nitrate (Fe(NO3)3·9H2O), Folic Acid, Glycine, HEPES, L-Arginine hydrochloride, L-Cystine 2HCl, L-Glutamine, L-Histidine hydrochloride-H2O, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Methionine, L-Phenylalanine, L-Serine, L-Threonine, L-Tryptophan, L-Tyrosine disodium salt dihydrate, L-Valine, Magnesium Sulfate (MgSO4) (anhyd.), Niacinamide, Phenol Red, Potassium Chloride (KCl), Pyridoxine hydrochloride, Riboflavin, Sodium Bicarbonate (NaHCO3), Sodium Chloride (NaCl), Sodium Phosphate monobasic (NaH2PO4-H2O), Thiamine hydrochloride, i-Inositol
10L
High Glucose, L-glutamine, HEPES, Phenol Red
Liquid
DMEM media
Storage: 2-8° C. Protect from light
Sodium Pyruvate

Safety and Handling

ShelfLife : 12 months from the date of manufacture

Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit