Learn More
Gibco™ DPBS with Calcium, Magnesium, Glucose and Pyruvate Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Balanced salt solution used for variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents
Manufacturer:  Gibco™ 14287072
 View more versions of this product
Catalog No. 14-287-072
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1000mL
No phenol red, sodium pyruvate
Calcium Chloride (CaCl2) (anhyd.), D-Glucose (Dextrose), Magnesium Chloride (MgCl2-6H2O), Potassium Chloride (KCl), Potassium Phosphate monobasic (KH2PO4), Sodium Chloride (NaCl), Sodium Phosphate dibasic (Na2HPO4-7H2O), Sodium Pyruvate
1X
Liquid
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.