DPBS with Calcium and Magnesium Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

For use with for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents

Manufacturer: Gibco 14040141

 View more versions of this product

Catalog No. 14-040-141

  Check Availability
Add to Cart

    Specifications

    Additive No phenol red, no sodium pyruvate
    Format Liquid
    Formulation Calcium Chloride (CaCl2) (anhyd.), Magnesium Chloride (MgCl2-6H2O), Potassium Chloride (KCl), Potassium Phosphate monobasic (KH2PO4), Sodium Chloride (NaCl), Sodium Phosphate dibasic (Na2HPO4-7H2O)
    Osmolality Osmality: 270 - 300mOsm/kg
    Concentration 1X
    Quantity 100mL
    pH pH range: 7.0 - 7.2,pH range: 7.0 - 7.2

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit