Fetal Bovine Serum, Certified, US Origin, Standard (Sterile-Filtered) Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Secure sera for cell culture with least viral risk sera from BSE-free regions

Manufacturer: Gibco 16000044

 View more versions of this product

Catalog No. 16-000-044

  Check Availability
Add to Cart

Disclaimers:

For In Vitro Diagnostic Use.


    Specifications

    Grade Certified
    Format Bottle
    Hemoglobin Concentration ≤ 10 mg/dL
    Quantity 500mL
    Endotoxin Concentration ≤ 5 EU/mL
    Heat Inactivated No

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit