Learn More
Fetal Bovine Serum, embryonic stem cell-Qualified, US Origin, Standard (Sterile-Filtered) Non-distribution product as customer accommodation.
Secure sera for cell culture with least viral risk sera from BSE-free regions
Manufacturer:  Gibco 16141061
 View more versions of this product
Catalog No. 16-141-061
  Check Availability
Add to cart
Disclaimers:
For In Vitro Diagnostic Use.
Specifications Description & Specifications
Specifications
≤50 EU/mL (levels routinely ≤ 10 EU/mL)
ES Cell-Qualified
≤25 mg/dL
No
Bottle
100mL