Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions

General stem cell culture media and reagents; sera for specialty research and specific assays

Supplier:  Gibco™ A3840202

Encompass

 View more versions of this product

Catalog No. A3840202


Add to cart

Description

Description

One-Shot 50-mL bottle features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500-mL bottles

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Triple Filtered at 0.1 µm
10 x 50 mL
10 x 50 mL
Sterile
One Shot™
Frozen
Fetal Bovine Serum, heat inactivated, qualified, One Shot™, USDA-approved regions
One Shot
Fetal Bovine Serum
Heat-Inactivated
Bovine
Triple Filtered, 0.1 µm
Qualified
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For In Vitro Diagnostic Use.