Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Fetal Bovine Serum, qualified, United States

Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Supplier:  Gibco™ A3160502

Encompass_Preferred

 View more versions of this product

Catalog No. A3160502


Add to Cart

Description

Description

If you’re one of the more than 80% of researchers who aliquots their FBS into 50 mL conical tubes, your life is about to change for the better. Transferring your FBS takes away your control — of how you spend your time, your contamination risk, and your volume consistency. It also wastes costly material.
The new Gibco™ One Shot™ FBS 50mL bottle design eliminates the need to aliquot, and the variability that comes with it. And that improves your chances for consistent results.

Product Features:
  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label has all the information you need
  • Generates 33% less waste than aliquoting from 500mL bottles

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Bovine
Liquid
Fetal Bovine Serum
Standard (Sterile-filtered)
Fewer resources used and less waste
One Shot™
Fetal Bovine Serum, qualified, One Shot™ format, United States
United States
One Shot
10 x 50 mL
Sterile
Qualified
Frozen
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit