Learn More
Gibco™ Human Flt-3 Ligand (FLT3L) Recombinant Protein
Fms-Like Tyrosine Kinase 3 Ligand (FLT3 Ligand, FLT3LG) is a bioactive protein intended for use in cell culture applications
$436.00 - $1578.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 35 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | FLT-3 Ligand |
Species | Human |
Description
Human Flt-3 Ligand (FLT3L) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
FLT3 Ligand is a cytokine involved in hematopoeisis.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human OCI-AML5 cells
Specifications
Cell Culture | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Human | |
Mammalian | |
Recombinant, Ligand | |
FLT-3 Ligand | |
Gibco™ |
35 kDa | |
FLT-3 Ligand | |
Lyophilized Human Flt-3 Ligand should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Flt-3 Ligand at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum. | |
< 0.1 ng/μg | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Signal Transduction, Stem Cell Research, Oncology | |
>95% by SDS-PAGE, >95% by SDS-PAGE |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.