Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ FreeStyle™ CHO Expression Medium
GREENER_CHOICE

Animal origin-free, chemically defined, protein-free medium

Supplier:  Gibco™ 12651014

Encompass_Preferred

 View more versions of this product

Catalog No. 12-651-014

  • $139.65 / Each of 1

    Save 10%

    Reg : $155.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Description

Description

FreeStyle CHO Expression Medium is specifically developed to support the growth and transient transfection of CHO-S cells in suspension without adaptation. This medium requires supplementation with 8 mM glutamine before use.

  • Formulation without hypoxanthine and thymidine for use with or without dhfr systems
  • Ability to support high-density suspension growth and transient transfection of CHO-S cells
  • Animal origin-free, protein-free and chemically defined formulation
  • Easy use as part of the FreeStyle MAX CHO Expression System for protein production
  • Scalability in spinner flasks and bioreactors

Formulation without hypoxanthine and thymidine for use with or without dhfr systems

  • Gibco FreeStyle CHO Expression Medium is made without hypoxanthine and thymidine for use in dihydrofolate reductase (dhfr) amplification systems
  • Non-dhfr systems require the addition of HT Supplement before use
Ability to support high-density suspension growth and transient transfection of CHO-S cells
  • Gibco FreeStyle CHO cells in suspension can be cultured directly into Gibco FreeStyle CHO Expression Medium without adaptation
  • Cells adapted to other commercially available serum-free media can be subcultured directly into Gibco FreeStyle CHO Expression Medium, usually without any further adaptation
  • Cells usually require adaptation from serum-containing formulations
  • Cells cultured in Gibco FreeStyle CHO Expression Medium can be transfected using lipid-based transfection reagents, such as FreeStyle MAX reagent
Animal origin-free, protein-free and chemically defined formulation
  • Gibco FreeStyle CHO Expression Medium is animal origin–free, protein-free, and chemically defined, allowing for easier purification of your protein of interest
  • Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition
Scalability in spinner flasks and bioreactors
  • Protein production using Gibco FreeStyle CHO Expression Medium can be scaled up in spinner flasks or bioreactors
  • The appropriate spinner or impeller speed and seeding density should be optimized for each system
  • Depending on the impeller design and speed, it may be necessary to supplement Gibco FreeStyle CHO Expression Medium with additional Pluronic™ F-68 to avoid sheer stress in the culture

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room temperature

Compliance

Gibco FreeStyle CHO Expression Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Expression Medium
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Suspension Cell Culture
No Glutamine
Serum-Free
Sustainable packaging
1000 mL
FreeStyle™ CHO-S Cells, CHO
Chemically Defined, Protein-Free, Serum-Free
≤0.25 EU/mL, ≤ 0.25 EU/ml
No Phenol Red
Room Temperature
Freestyle™, Gibco™
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.