Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ GM-CSF Recombinant Swine Protein Non-distribution product as customer accommodation.

Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications

Specifications

Purity 95 %
Purification Method Sequential Chromatography
Gene Alias GM-CSF
Gene ID (Entrez) 397208
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PSC2014 Non-distribution product as customer accommodation.
View Documents
Gibco™
PSC2014
10μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Recombinant swine GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.

Approved for shipment on Wet or Dry Ice.

Specifications

Specifications

95 %
Sequential Chromatography
GM-CSF
397208
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit