Learn More
Gibco™ Goat Serum, New Zealand origin
Excellent value for basic cell culture, specialty research, and specific assays
$68.65 - $251.65
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Adult |
Species | Goat/Caprine |
Country of Origin | New Zealand |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-210-064
|
Gibco™
16210064 |
100 mL |
Each
for $68.65
Save 6% Reg : |
|
|||||
16-210-072
|
Gibco™
16210072 |
500 mL |
Each
for $251.65
Save 6% Reg : |
|
|||||
Description
- Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support
- Helps meet research needs and budget requirements
- Shipped frozen, store at <-10°C
- Sera Category: Specialty
- Origin: New Zealand
- Age: Adult
Order Info
Shipping Condition: Dry ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Goat/Caprine | |
Liquid | |
Standard (Sterile-filtered) | |
Sustainable packaging | |
Frozen |
Adult | |
New Zealand | |
Goat Serum | |
Sterile | |
Gibco™ |
For In Vitro Diagnostic Use. For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.