Hybridoma-SFM Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Specifically developed to support the growth of hybridomas for antibody production

Manufacturer: Gibco 12045076

 View more versions of this product

Catalog No. 12-045-076

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Format Liquid
    For Use With (Application) Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
    Quantity 1000mL
    Storage Requirements Storage conditions: 2°C to 8°C. Protect from light. Shelf life: 12 months from date of manufacture
    Additive L-Glutamine, phenol red

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit