Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ IL8 Monocyte Recombinant Human Protein Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications

$267.00 - $3960.00

Specifications

Gene Alias IL-8 Monocyte
Gene ID (Entrez) 3576
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PHC0884 Non-distribution product as customer accommodation.
View Documents
Gibco™
PHC0884
25μg
Each for $267.00
Add to cart
 
PHC0885 Non-distribution product as customer accommodation.
View Documents
Gibco™
PHC0885
50μg
Each for $500.00
Add to cart
 
PHC0883 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Gibco™
PHC0883
1mg
Each for $3,960.00
Add to cart
 
Description

Description

IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.

Shipping Condition: Wet Ice.

Specifications

Specifications

IL-8 Monocyte
3576
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit