IMDM, Powder Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Iscove's Modified Dulbecco's Medium is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells

Manufacturer: Gibco 12200036

 View more versions of this product

Catalog No. 12-200-036

  Check Availability
Add to Cart

Disclaimers:

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.


Required Accessories : Supplementation

    Specifications

    Formulation Biotin, Calcium Chloride (CaCl2) (anhyd.), Choline chloride, D-Calcium pantothenate, D-Glucose (Dextrose), Folic Acid, Glycine, HEPES, L-Alanine, L-Arginine hydrochloride, L-Asparagine (freebase), L-Aspartic acid, L-Cystine 2HCl, L-Glutamic Acid, L-Glutamine, L-Histidine hydrochloride-H2O, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tryptophan, L-Tyrosine disodium salt, L-Valine, Magnesium Sulfate (MgSO4) (anhyd.), Niacinamide, Phenol Red, Potassium Chloride (KCl), Potassium Nitrate (KNO3), Pyridoxal hydrochloride, Riboflavin, Sodium Chloride (NaCl), Sodium Phosphate monobasic (NaH2PO4-H2O), Sodium Pyruvate, Sodium Selenite (Na2SeO3-5H20), Thiamine hydrochloride, Vitamin B12, i-Inositol
    Format Powder
    Quantity 10 x 1L,10 x 1L
    Storage Requirements Storage: 2-8° C. Shelf life: 36 months from date of manufacture
    With Additives Sodium Pyruvate; High Glucose; L-Glutamine; Phenol Red,Sodium Pyruvate; High Glucose; L-Glutamine; Phenol Red

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit