Learn More
Gibco™ Mycophenolic Acid Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Produced by Penicillium brevi-compactum, Penicillium loniferum and related species
Manufacturer:  Gibco™ 11814019
Catalog No. 11-814-019
Add to cart
Description
Description
Gibco™ Mycophenolic Acid blocks the guanosine monophosphate (GMP) synthesis pathway.
  • Available as 500mg powder
  • Used as selective agent in mammalian systems, commonly at a concentration of 25µg/mL
Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Mycophenolic Acid
Powder
500mg
Antibiotic (Selective)
Eukaryotic Selection/Stable Cell Line Generation
Mycophenolic Acid
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.