Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ PDGF-BB Recombinant Mouse Protein Non-distribution product as customer accommodation.

Recombinant Mouse Platelet Derived Growth Factor-BB (PDGF-BB) is a bioactive protein intended for use in cell culture applications

$277.00 - $497.00

Specifications

Purity 95 %
Purification Method Sequential Chromatography
Gene Alias PDGF-BB
Gene ID (Entrez) 18591
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PMG0044 Non-distribution product as customer accommodation.
View Documents
Gibco™
PMG0044
10μg
Each for $277.00
Add to cart
 
PMG0045 Non-distribution product as customer accommodation.
View Documents
Gibco™
PMG0045
25μg
Each for $497.00
Add to cart
 
Description

Description

Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose-dependent proliferation of BALB/c 3T3 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

95 %
Sequential Chromatography
PDGF-BB
18591
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit