Sf-900™ III SFM Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Low-hydrolysate, serum-free, protein-free, animal origin-free

Manufacturer: Gibco 12658019

 View more versions of this product

Catalog No. 12-658-019

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Format Liquid
    Species S. frugiperda, Spodoptera frugiperda
    For Use With (Application) Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
    Quantity 500mL
    Additive L-Glutamine
    Cell Line Sf21, Sf9
    Storage Requirements Storage conditions: 2° to 8°C. Protect from light. Shelf life: 18 months from date of manufacture

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit