Gibson Bioscience Campy Gas Supplies
Supplies for Campylobacter gas generator system
Manufacturer: GIBSON LABORATORIES LLC 100125
 View more versions of this product
Catalog No. 21-200-498
  Check Availability
Add to Cart
    Specifications
    Product Type Cylinder
    Size 1230 mult.
    Includes Gas (5% oxygen, 10% carbon dioxide and 85% nitrogen)
    Disposable Yes
    For Use With (Application) Campylobacter incubation
  • Description & Specifications
Related Products
Product Content Feedback