Learn More
Gibson Bioscience Reagents: Kovacs
For the detection of microorganisms that produce Indole as a by-product of their metabolism
Manufacturer:  GIBSON LABORATORIES LLC 60017
 View more versions of this product
Catalog No. 21-200-317
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Detect those microorganisms which produce Indole as a by-product of their metabolism
50mL
18 to 24 hr.
Kovacs Reagent
Isoamyl alcohol 750mL, hydrochloric acid 250mL, p-Dimethylaminobenzaldehyde 50g
2° to 25°C
50mL