Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Aqueous IFD, 0.2µm, 50mm, nylon Available on GSA/VA Contract for Federal Government customers only.

Aqueous IFD from GE is a polypropylene housed in-line filter/degasser (IFD) with nylon membrane. It connects to an HPLC line and simultaneously filters and degasses during HPLC mobile phase preparation.

$240.00 - $261.50

Specifications

Pore Size 0.2μm
No. per Pack 10
Housing Material Polypropylene (PP)
For Use With (Application) Whatman Aqueous and Solvent IFD Inline Filter Degassers
Filtration Area (Metric) 16 cm2
View More Specs

Products 2
Catalog Number Mfr. No. Inlet Connection (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Inlet Connection (Metric) Price Quantity & Availability  
09-703-90 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67265002
0.8 to 4 mm tubing Pack of 10 for $240.00
Add to cart
 
05-713-407
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
67265002A
1/8 in. tubing only Pack of 10 for $261.50
Add to cart
 
Description

Description

  • Faster than traditional methods of HPLC mobile phase preparation, saving time in the laboratory
  • Provides pure filtration of aqueous mobile phases without expensive degassing equipment
  • Chemically resistant polypropylene construction enables reliable filtration
  • Can be integrity-tested with the bubble point method
  • Includes end fittings – for replacements, see our IFD end fitting kit

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0.2μm
Polypropylene (PP)
16 cm2
10
Whatman Aqueous and Solvent IFD Inline Filter Degassers
Aqueous IFD from GE is a polypropylene housed in-line filter/degasser (IFD) with nylon membrane. It connects to an HPLC line and simultaneously filters and degasses during HPLC mobile phase preparation.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit