Learn More
GIGYF2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 247901AP
Catalog No. 50-173-4545
Add to cart
Description
Description
The function of this protein remains unknown.
Specifications
Specifications
GIGYF2
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6Y7W6
GIGYF2, GYF2, KIAA0642, PARK11, PERQ2, PERQ3, TNRC15
Rabbit
IgG
150 μL
-20°C
Primary
26058
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
GIGYF2
Liquid
GIGYF2
GIGYF2 Fusion Protein Ag20329
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human