Learn More
Masterflex™ Variable-Area Flowmeter, Correlated Reading
Don’t sacrifice suitability for precision.
 View more versions of this product
Catalog No. 15-078-102
Add to cart
Description
Description
Thermo Scientific™ Gilmont Laboratory Correlated Flowmeters have excellent chemical compatibility, with glass and PTFE construction. High-accuracy correlated flowmeters are available in a wide selection of flow ranges.
Extremely accurate - ±2% of reading or ±1 scale division, whichever is greater. Flowmeters with micro-sized tubes (models 03210-00, 03210-20, and 03234-50) are accurate to ±5% of reading or ±2% of scale whichever is greater. Computer calibrated flow charts are included for floats used with both air and water at STP. An “R factor” chart and formulas are included to convert scale readings for other gases or liquids or for floats other than glass. Order Cole-Parmer's flow analysis software below to generate flow charts specific to your individual applications.
Shielded models accept 3/8 in. ID tubing (sizes 4 and 5 accept 5/8 in. ID tubing). PTFE coupling adapters for unshielded models are not included—order separately.
Specifications
Specifications
0.02 to 15mL/min.
-26° to +65°C (-15° to +150°F) at full pressure rating. Above 65°C (150°F), max. pressure is 40psig
Ruby
Use taper joint adapters
Unshielded; Tube size: micro
0.0002 to 0.12mL/min
15psi
±5% of reading or ±2 scale division (Air) ±10% of
Viton™
Unshielded
Micro
±1% of reading or ±0.5 scale division, whichever is greater