Learn More
Masterflex™ Variable-Area Flowmeter, Correlated Reading
Don’t sacrifice suitability for precision.
$420.50 - $2200.00
Specifications
Repeatability ±1% of reading or ±0.5 scale division, whichever is greater
Seal Material Viton™
Temperature Rating -26° to +65°C (-15° to +150°F) at full pressure rating. Above 65°C (150°F), max. pressure is 40psig
View More Specs
Products 15
Catalog Number Description Airflow Rate Price Quantity & Availability  
Catalog Number Description Airflow Rate Price Quantity & Availability  
15-078-111
View Documents
Unshielded 10 to 1,900mL/min. (Glass); 20 to 3,400mL/min. (SS) Each for $600.00
Add to cart
15-078-107
View Documents
Shielded 0.2 to 100 mL/min. (Glass); 0.36 to 180 mL/min. (SS) Each for $1,279.00
Add to cart
15-078-110
View Documents
Shielded w/valves 1 to 280mL/min. (Glass); 2 to 500mL/min. (SS) Each for $1,326.00
Add to cart
15-078-105
View Documents
Unshielded 0.2 to 100mL/min. (Glass); 0.36 to 180mL/min. (SS) Each for $544.00
Add to cart
15-078-104
View Documents
Shielded w/valves 0.02 to 15mL/min. Each for $1,215.00
Add to cart
15-078-102
View Documents
Unshielded 0.02 to 15mL/min. Each for $420.50
Add to cart
15-078-113
View Documents
Shielded w/valves 10 to 1,900mL/min. (Glass); 20 to 3,400mL/min. (SS) Each for $1,345.00
Add to cart
15-078-116
View Documents
Shielded w/valves 200 to 14,000mL/min. (Glass); 360 to 25,000mL/min. (SS) Each for $1,369.00
Add to cart
15-078-117
View Documents
Unshielded 1000 to 36,000mL/min. (Glass); 1800 to 64,000mL/min. (SS) Each for $570.50
Add to cart
15-078-120
View Documents
Unshielded 3000 to 77,000mL/min. (Glass); 5300 to 137,000mL/min. (SS) Each for $648.50
Add to cart
15-078-118
View Documents
Shielded 1000 to 36,000mL/min. (Glass); 1800 to 64,000mL/min. (SS) Each for $869.50
Add to cart
15-078-135
View Documents
Shielded 25,000 to 330,000mL/min. (Glass); 50,000 to 675,000mL/min. (SS) Each for $1,579.00
Add to cart
15-078-122
View Documents
Shielded w/valves 3000 to 77,000mL/min. (Glass); 5300 to 137,000mL/min. (SS) Each for $2,200.00
Add to cart
15-078-121
View Documents
Shielded 3000 to 77,000mL/min. (Glass); 5300 to 137,000mL/min. (SS) Each for $1,009.00
Add to cart
15-078-114
View Documents
Unshielded 200 to 14,000mL/min. (Glass); 360 to 25,000mL/min. (SS) Each for $525.50
Add to cart
Description
Description
Thermo Scientific™ Gilmont Laboratory Correlated Flowmeters have excellent chemical compatibility, with glass and PTFE construction. High-accuracy correlated flowmeters are available in a wide selection of flow ranges.
Extremely accurate - ±2% of reading or ±1 scale division, whichever is greater. Flowmeters with micro-sized tubes (models 03210-00, 03210-20, and 03234-50) are accurate to ±5% of reading or ±2% of scale whichever is greater. Computer calibrated flow charts are included for floats used with both air and water at STP. An “R factor” chart and formulas are included to convert scale readings for other gases or liquids or for floats other than glass. Order Cole-Parmer's flow analysis software below to generate flow charts specific to your individual applications.
Shielded models accept 3/8 in. ID tubing (sizes 4 and 5 accept 5/8 in. ID tubing). PTFE coupling adapters for unshielded models are not included—order separately.
Specifications
Specifications
±1% of reading or ±0.5 scale division, whichever is greater
Viton™
-26° to +65°C (-15° to +150°F) at full pressure rating. Above 65°C (150°F), max. pressure is 40psig