Learn More
Masterflex™ Variable-Area Flowmeter with Valve
Design makes each meter suitable for a broad range of laboratory applications.
 View more versions of this product
Catalog No. 15-078-123
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Thermo Scientific™ Gilmont laboratory direct-reading flowmeters have excellent chemical compatibility with glass and PTFE construction. Flowmeter measurements can be read directly off the flowmeter.
Choose from unshielded, shielded, or shielded with valve flow meters suitable for a wide range of applications.
Unshielded Flowmeters work well in low-pressure applications. Connect the flowtube end directly to tubing having the proper inner diameter. Maximum pressure rating for unshielded flowmeters is 15 psi.
Shielded Flowmeters are better suited to higher-pressure applications or for installations requiring a panel-mounted flowmeter. End bushings are molded with dual connection capability—tubing or threaded.
Shielded Flowmeters with Valves offer flow monitoring along with precise control through an integrated 20-turn micrometer valve. The valve can be adjusted from 0.1 to 100% of maximum flow (semilogarithmic) and provides recise regulation from 0.3 to 60% of maximum flow.
Specifications
Specifications
Polycarbonate
15psi
±5% of reading or 2mm of scale length, whichever is greater
Unshielded
10
±1% of reading or ±0.5 scale division, whichever is greater
0.2 to 90mL/min. (Glass); 0.36 to 160mL/min. (SS)
-26°C to +65°C (-15°F to +150°F) at full pressure rating. Above 65°C (150°F), max. pressure is 40psig
Use taper joint adapters
Unshielded; Tube size: 10
0.002 to 1.1mL/min. (Glass); 0.004 to 2.3mL/min. (SS)