Learn More
Masterflex™ Variable-Area Flowmeter with Valve
Design makes each meter suitable for a broad range of laboratory applications.
$298.50 - $683.00
Specifications
Case Material Polycarbonate
Repeatability ±1% of reading or ±0.5 scale division, whichever is greater
Temperature Rating -26°C to +65°C (-15°F to +150°F) at full pressure rating. Above 65°C (150°F), max. pressure is 40psig
Accuracy ±5% of reading or 2mm of scale length, whichever is greater
View More Specs
Products 5
Catalog Number Description Tube Size Price Quantity & Availability  
Catalog Number Description Tube Size Price Quantity & Availability  
15-078-133
View Documents
Unshielded 15 Each for $457.50
Add to cart
15-078-134
View Documents
Shielded 15 Each for $683.00
Add to cart
15-078-123
View Documents
Unshielded 10 Each for $378.00
Add to cart
15-078-129
View Documents
Unshielded 13 Each for $298.50
Add to cart
15-078-127
View Documents
Unshielded 12 Each for $348.00
Add to cart
Description
Description
Thermo Scientific™ Gilmont laboratory direct-reading flowmeters have excellent chemical compatibility with glass and PTFE construction. Flowmeter measurements can be read directly off the flowmeter.
Choose from unshielded, shielded, or shielded with valve flow meters suitable for a wide range of applications.
Unshielded Flowmeters work well in low-pressure applications. Connect the flowtube end directly to tubing having the proper inner diameter. Maximum pressure rating for unshielded flowmeters is 15 psi.
Shielded Flowmeters are better suited to higher-pressure applications or for installations requiring a panel-mounted flowmeter. End bushings are molded with dual connection capability—tubing or threaded.
Shielded Flowmeters with Valves offer flow monitoring along with precise control through an integrated 20-turn micrometer valve. The valve can be adjusted from 0.1 to 100% of maximum flow (semilogarithmic) and provides recise regulation from 0.3 to 60% of maximum flow.
Specifications
Specifications
Polycarbonate
±1% of reading or ±0.5 scale division, whichever is greater
-26°C to +65°C (-15°F to +150°F) at full pressure rating. Above 65°C (150°F), max. pressure is 40psig
±5% of reading or 2mm of scale length, whichever is greater