Learn More
Gilmont™ Micrometer Syringes and Burets
Experience high precision with economy of cost when using Gilmont™ Micrometer Syringes.
 View more versions of this product
Catalog No. 15-078-153
  Check Availability
Add to cart
Specifications
Description
Experience high precision with economy of cost when using Thermo Scientific™ Gilmont Micrometer Syringes.
  • 0.5% accuracy
  • Liquid comes in contact with only glass, VITON™ and PTFE™
  • VITON O-ring gives a vacuum tight seal
  • All other plastic parts of polypropylene--instrument may be autoclaved
  • Available in two sizes, 0.2mL  in 0.0002mL div. and 2.0mL in 0.002mL div.
  • Fills and delivers liquid with micrometer control
  • Simple to clean and assemble
Description & Specifications
Specifications
Buret
2.0mL