Learn More
Gilson™ EXPERT™ Universal Fit Pipette Filter Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
Manufacturer:  Gilson™ F1735031
 View more versions of this product
Catalog No. F1735031G
Add to cart
Description
Description
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
Polypropylene
Sterilized filter tips
Clear
5.857cm
10, 50, 100μL
1 to 200 μL
EF200ST