Learn More
Gilson™ EXPERT™ Universal Fit Pipette Filter Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
$71.20 - $144.50
Specifications
Description Sterilized filter tips
View More Specs
Products 8
Catalog Number Mfr. No. Graduations Model Length (Metric) Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Graduations Model Length (Metric) Volume (Metric) Price Quantity & Availability  
F1731031G
View Documents
Gilson™
F1731031
2 and 10μL EF10ST 2.883cm 0.1 to 10 μL
Pack of 960 for $144.50
Add to cart
F1732031G
View Documents
Gilson™
F1732031
2, 5, 10μL EFL10ST 4.534cm 0.1 to 10 μL
Pack of 768 for $144.50
Add to cart
F1733031G
View Documents
Gilson™
F1733031
10, 50, 100μL EF20ST 5.144cm 1 to 20 μL
Pack of 768 for $144.50
Add to cart
F1734031G
View Documents
Gilson™
F1734031
10, 50, 100μL EF100ST 5.144cm 1 to 100 μL
Pack of 768 for $144.50
Add to cart
F1735031G
View Documents
Gilson™
F1735031
10, 50, 100μL EF200ST 5.857cm 1 to 200 μL
Pack of 768 for $144.50
Add to cart
F1735041G
View Documents
Gilson™
F1735041
10, 50, 100μL Tri-Fit EF200ST XL Extended Length 7.75cm 1 to 200 μL
Pack of 384 for $71.20
Add to cart
F1736031G
View Documents
Gilson™
F1736031
10, 50, 100μL EF300ST XL Extended Length 7.75cm 1 to 300 μL
Pack of 384 for $71.20
Add to cart
F1737031G
View Documents
Gilson™
F1737031
200, 500, 1000μL EF1000ST XL Extended Length 10.22cm 50 to 1250 μL
Pack of 384 for $71.20
Add to cart
Description
Description
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
Sterilized filter tips