Learn More
Gilson™ EXPERT™ Universal Fit Standard Pipette Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
Manufacturer:  Gilson™ F1735011
 View more versions of this product
Catalog No. F1735011G
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
Standard Pipette Tips
200, 500, 1000μL
10.22cm
50 to 1250μL
E1000ST XL Sterilized Tipack