Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ EXPERT™ Universal TOWERPACK™ Racked Pipette Tips Reload Kit

Be green, save space: snaps tips into place in seconds with just one hand with tips plus Tipack boxes

Manufacturer:  Gilson™ F1732061

 View more versions of this product

Catalog No. F1732061G


Add to cart

Description

Description

Gives you easy access to 10 layers of tips that easily snap into the Tipack boxes.

  • Fit leading pipetter brands
  • Allow you to stock one brand of tip with practical packaging options so you can save time, money and bench space
  • Produced in a sterile, clean-room environment using the purest raw materials and state-of-the-art manufacturing
  • Offer consistent performance
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and pyrogens.
  • Feature an inert polymer hydrophobic surface to minimize sample binding when dispensing
  • Clear packaging allows easy tip identification
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipetters
  • Feature graduations at varying volumes allowing for quick checks when aspirating samples
  • Economical and easy to use

  • TowerPack Kits: 960 Tips/Pk., 10 empty Tipack boxes
  • E1000 XL TowerPack Kit: 768 Tips/Pk., 8 empty Tipack Boxes

Specifications

Specifications

Universal Fit
0.1 to 10μL
Red
EL10
1.78 in.
DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free
Universal Fit Pipettor
Non-sterile
2, 5, 10μL
Inert Polymer
4.53cm
Universal Fit Pipettor
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit