Learn More
Gilson™ MICROMAN™ Capillary Pistons Available on GSA/VA Contract for Federal Government customers only.
Made specifically for Gilson™ MICROMAN™ E pipets.Gilson™ MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs perfectly.
Manufacturer:  Gilson™ F148315
 View more versions of this product
Catalog No. F148315G
Add to cart
Description
Description
  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs perfectly.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Gilson™ Microman™
10 to 100μL
CP100ST
Gilson™ Microman™
Box of 960
Sterile
PP (Polypropylene)