Learn More
Gilson™ PIPETMAN™ EasyPack™ Tips
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMAN™ EasyPack™ Tips.
Supplier:  Gilson™ F161451
 View more versions of this product
Catalog No. F161451G
Add to Cart
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
Specifications
Specifications
0.1 to 20 μL
Non-sterile
Autoclavable
12 cm
Universal
Pipet Tips
Reusable
Pipetman EcoPack
Non-filtered
Clear
4.72 in.
Bagged
5 Bags of 200 Tips
Red
Yes
Polypropylene
Product Suggestions
Product Suggestions