Learn More
Gilson™ PIPETMAN™ EasyPack™ Tips Available on GSA/VA Contract for Federal Government customers only.
Sealed with an easy open/close zipper lock. Gilson™ PIPETMAN™ EasyPack™ Tips come in bulk packs with assorted sizes.
Manufacturer:  Gilson™ F161671
 View more versions of this product
Catalog No. F161671G
Add to cart
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface
  • Manufactured in a cleanroom and hence no risk of contamination
  • Full traceability from the mold to the bench guaranteed
  • Volume graduations make it easy to visually check the volume aspirated
  • Requires low tip-mounting force
  • Easy identification with color-coded packaging
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5 bags of 200 tips
100 to 1000μL
PP (Polypropylene)
Universal Fit
Yes