Learn More
Gilson™ PIPETMAN™ EasyPack™ Tips
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMAN™ EasyPack™ Tips.
Supplier:  Gilson™ F161730/1
 View more versions of this product
Catalog No. F1617301G
Add to Cart
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
Specifications
Specifications
20 to 300 μL
Non-sterile
5.63 in.
Bagged
Bag of 1000 Tips
Yes
Polypropylene
Non-filtered
Autoclavable
14.3 cm
Universal
Pipet Tips
Pipetman EasyPack
Product Suggestions
Product Suggestions