Learn More
Gilson™ PIPETMAN™ Diamond Blister Pack Refill Pipette Tips
Autoclavable or gamma-ray sterlized
Manufacturer:  Gilson™ F172210
 View more versions of this product
Catalog No. F172210G
Add to cart
Description
Description
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Standard or filtered tips
  • Validated for use with 384-well tips: F172211G, F172311G and F17511G
Specifications
Specifications
Universal Fit
0.1 to 20μL
Non-sterile
DSL10
Blister Refill
Non-filtered
Yes
Autoclavable