Learn More
Gilson™ PIPETMAN™ EXPERT™ Sterilized Filter Pipette Tips
Features graduations at varying volumes allowing for quick-checks when aspirating samples
Manufacturer:  Gilson™ F1735032
 View more versions of this product
Catalog No. F1735032G
Add to cart
Description
Description
  • Features patented, low-retention polymer that creates hydrophobic surface to minimize sample binding when dispensing
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
1 to 200μL
Sterile
10, 50, 100μL
Filtered
Clear
5.857cm