Learn More
Gilson™ PIPETMAN™ EXPERT™ Universal Fit Gel Loading Pipet Tips: Sterile
Flat or round tip styles are available
Manufacturer:  Gilson™ F1733382
Alternative option to part number 53550190
 View more versions of this product
Catalog No. F1733382G
Add to cart
Description
Description
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
Specifications
Specifications
Flat
Sterile
Flat
1 to 200μL
5μL
8.255cm