Learn More
Gilson™ PIPETMAN™ F Fixed Volume Pipettes Available on GSA/VA Contract for Federal Government customers only.
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer:  Gilson™ F123605
 View more versions of this product
Catalog No. F123605G
Add to cart
Description
Description
  • Dedicated fixed-volume pipette assures reliable and consistant results
  • Volume can be adjusted by users in the laboratory for dense or viscous fluids
  • Single-volume mechanism provides a low-cost alternative for clinical diagnostics, quality control and routine testing
Gilson maximum permissible errors are guaranteed only when PIPETMAN pipettes are used with the recommended Gilson PIPETMAN Tips.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
200μL
F200
GLP
D200, DF100ST, DF200ST, DF300ST tip
Lifetime
±1.6μL
D200, DF100ST, DF200ST, DF300ST tip
Stainless Steel piston and PVDF handle