Learn More
Gilson™ PIPETMAN™ G Pipets Available on GSA/VA Contract for Federal Government customers only.
Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.
Manufacturer:  Gilson™ F144056M
 View more versions of this product
Catalog No. F144056MG
Add to cart
Description
Description
  • Body (handle) in PVDF minimizes the hand-warming effect during pipetting and is resistant against agressive chemicals
  • Direct volume reading in decimal digits helps prevent mistakes in volume setting
  • Comes with classic metal or plastic tip ejector
  • Clip ejector fits securely into the body of the pipette and helps avoid slipping over time
  • Redesigned seal, lighter ejection springs and high quality piston grease offer as low as 2 Newtons of force to pipet
  • High quality piston grease
Model Specific Features
  • Models P2G and P10G incorporate dual position tip ejectors for use with D10 (short) and DL10 (long) tips
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Trade in program available. Please contact your Fisher Scientific sales representative for more details.
Specifications
Specifications
±0.1, ±0.1, ±0.2μL
Extremely low pipetteting forces, lightweight design
Stainless Steel tip ejector
Yellow
2 to 20μL
D200, DF30ST tip
D200, DF30ST tip
Stainless Steel and PVDF
P20G