Learn More
Gilson™ PIPETMAN™ L Mechanical Pipets
Combines the legendary robustness of the PIPETMAN Classic™ with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Gilson™ PIPETMAN™ L Mechanical Pipets are available in 8 single channel models covering the volume range of 0.2µL to 10mL.
Manufacturer:  Gilson™ FA10006P
 View more versions of this product
Catalog No. FA10006PG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mechanical pipette combines the legendary robustness of the PIPETMAN™ Classic with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Available in 8 single channel models covering the volume range of 0.2µL to 10mL.
  • Redesigned handle fits naturally in user's hand, no matter the size
  • Volume locking system guarantees no accidental change in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • 2D identification code allows easy tracking of product information
  • New piston assembly designed to drastically improve pipetting and purge forces
  • Redesigned tip ejector shape and internal mechanism offers reduced tip ejection forces by 50%
Order Info
Trade in program available, please contact your Fisher Scientific Sales representative for more details.
Specifications
Specifications
±3, ±4, ±8μL
Ergonomic
D1000, D1200, DF1000ST, DF1200ST tip
Blue
100 to 1000μL
D1000, D1200, DF1000ST, DF1200ST tip
Transporting a measured volume of liquid
Plastic tip ejector
P1000L