Gilson™ PIPETMAN™ M Multichannel Pipettes
Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
Manufacturer: Gilson™ F81007
Catalog No. F81007G
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Compatible Tips D200, D300, DF100ST, DF200ST, DF300ST tips
    Finger Style Ergonomic
    Warranty 1 Year
    Color Yellow,Yellow
    Model P8x200M
    Number of Channels 8
    Volume (Metric) 20 to 200µL
    Product Type Multichannel Pipette
    Compatibility D200, D300, DF100ST, DF200ST, DF300ST tips
    • Motorized piston with adjustable piston speed that is compatible with varying solution densities
    • Lockable volume to prevent inaccuracies
    • Color-coded push-button corresponds to matching tip packs
    • Service alert reminders
    • Unique tip holder design allows for a secure fit of most brands of tips
    • Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
    • Pipette can be used while charging eliminating bottlenecks and downtime
    • Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
    • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
    • Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
  • Description & Specifications
Related Products
Product Content Feedback