Learn More
Gilson™ PIPETMAN™ M Pipettes
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
Specifications
Product Line Pipetman™
Type Pipette
Finger Style Ergonomic
Warranty 1 Year
View More Specs
Products 2
Catalog Number Mfr. No. Number of Channels Model Volume (Metric) Compatible Tips Price Quantity & Availability  
Catalog Number Mfr. No. Number of Channels Model Volume (Metric) Compatible Tips Price Quantity & Availability  
F81015G
View Documents
Gilson™
F81015
1 P1200M 100 to 1200 μL D1000, DF1000, D1200, DF1200 Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
F81016G
View Documents
Gilson™
F81016
1 P5000M 500 to 5000 μL D5000 Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
  • Motorized piston with adjustable piston speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
  • Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
Specifications
Pipetman™
Pipette
Ergonomic
1 Year