Learn More
Gilson™ PIPETMAN M Single Channel Pipets
Fully motorized, lightweight pipet that virtually eliminates pipetting forces and minimizes RSI (Repetitive Strain Injuries) among users.
Manufacturer: Gilson™ F81013
Description
An ergonomic pipetting solution with GLP features that each lab needs
- Motorized piston with adjustable speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes – reverse, mix, repetitive and pipetting – to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
Specifications
5 to 100 μL | |
D1000, D1200, DF1000ST, DF1200ST pipette tip | |
Fully motorized piston, color coded push button, universal tip holder |
1 | |
P100M |