Learn More
Gilson™ PIPETMAN M Single Channel Pipets
Fully motorized, lightweight pipet that virtually eliminates pipetting forces and minimizes RSI (Repetitive Strain Injuries) among users.
Manufacturer:  Gilson™ F81024
 View more versions of this product
Catalog No. F81024G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
An ergonomic pipetting solution with GLP features that each lab needs
  • Motorized piston with adjustable speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes – reverse, mix, repetitive and pipetting – to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
Specifications
Specifications
20 to 200 μL
D200, D300, DF200ST, DF300ST pipette tip
Fully motorized piston, color coded push button, universal tip holder
1
P200M