Learn More
Gilson™ PIPETMAN M Single Channel Pipets
Fully motorized, lightweight pipet that virtually eliminates pipetting forces and minimizes RSI (Repetitive Strain Injuries) among users.
Specifications
Includes Fully motorized piston, color coded push button, universal tip holder
View More Specs
Products 4
Catalog Number Mfr. No. Model Volume (Metric) For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Model Volume (Metric) For Use With (Equipment) Price Quantity & Availability  
F81022G
View Documents
Gilson™
F81022
P10M 0.5 to 10 μL D10, DL10, DF10ST, DFL10ST pipette tip This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
F81023G
View Documents
Gilson™
F81023
P20M 2 to 20 μL D200, DF30ST pipette tip This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
F81013G
View Documents
Gilson™
F81013
P100M 5 to 100 μL D1000, D1200, DF1000ST, DF1200ST pipette tip This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
F81024G
View Documents
Gilson™
F81024
P200M 20 to 200 μL D200, D300, DF200ST, DF300ST pipette tip This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
An ergonomic pipetting solution with GLP features that each lab needs
  • Motorized piston with adjustable speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes – reverse, mix, repetitive and pipetting – to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
Specifications
Specifications
Fully motorized piston, color coded push button, universal tip holder