Learn More
Gilson™ PIPETMAN Neo™ Multichannel Pipets Available on GSA/VA Contract for Federal Government customers only.
Offers an excellent compromise between comfort and target precision. Gilson™ PIPETMAN Neo™ Multichannel Pipets are designed to apply equal pressure on each piston to ensure consistent volume levels across all channels.
Manufacturer:  Gilson™ F14402
 View more versions of this product
Catalog No. F14402G
Add to cart
Description
Description
  • Easy-to-remove tip ejector with autoclavable lower part
  • Internal design applies equal pressure on each piston to ensure consistent volume levels across all channels
  • PVDF handle minimizes the hand-warming effect during pipetting and is resistant to aggressive chemicals
  • Gilson Trident and unique spacer create a stepped ejection, staggering the tip ejection and thus reducing the force required to eject tips
  • Unique tip holder allows for a secure fit of most brands of tips
  • Available in 8- and 12-channel models
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2 to 20μL
P12x20N
Multichannel Pipet
1 Year
12