Learn More
Gilson™ PIPETMAN™ Neo Pipets Available on GSA/VA Contract for Federal Government customers only.
Fits most brands of tips. Gilson PIPETMAN™ Neo Pipets are the next generation of the PIPETMAN Classic™ with lower spring and ejection forces. 
Manufacturer:  Gilson™ F144563
 View more versions of this product
Catalog No. F144563G
Add to cart
Description
Description
  • Redesigned tip ejector and ejection springs reduces tip ejection forces
  • Unique clip ejector fits securely into the body of the pipette to avoid slipping over time
  • PVDF handle limits the transfer of heat from hand to the pipette and is resistant to aggressive chemicals
  • Stainless steel tip ejector resists corrosion and offers a smooth tip ejection stroke
  • Color-coded push-button for ease of identification
  • Unique tip holder allows for a secure fit of most brands of tip
  • Models P2N and P10N incorporate dual position tip ejectors, allowing the use of D10 (short) and DL10 (long)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
±0.1, ±0.1, ±0.2μL
Ergonomic
D200, DF30ST
1
ISO8655
Color coded push button, PVDF handle, Stainless Steel tip ejectors, universal tip holder
P20N
2 to 20μL